Learn More
Total Base Number Standards, Paragon Scientific™
Click to view available options
Concentration:
15.17 mg KOH/g
3.01 mg KOH/g
30.17 mg KOH/g
40.51 mg KOH/g
6.06 mg KOH/g
70.24 mg KOH/g
Description
Each standard is certified in strict accordance with international test method protocols, ASTM D2896 / IP 276. Manufacture and certification is carried out in strict accordance with Paragon Scientific’s dual UKAS accreditations to ISO 17025 & ISO 17034. This dual accreditation allows us to ensure the highest level of accreditation guarantee and provides the most credible certified data available worldwide
- Base Number (BN) tested and certified in accordance with ASTM D664 / IP 177
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
- Typically used in, but not limited to, the analysis of used oils and lubricants
- Supplied as single volume packs or as triple packs
- Supplied in tamper-evident glass packaging, providing assurance of sample integrity
- Manufactured in the United Kingdom
- 24 month shelf life
Specifications
Concentration | 40.51 mg KOH/g |
Quantity | 50 g |
Nominal values quoted