missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNPO3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 €
Specifications
| Antigen | TNPO3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TNPO3 Polyclonal specifically detects TNPO3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| TNPO3 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EAW83708 | |
| 23534 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human TNPO3. The immunogen is located within amino acids 220 - 270 of TNPO3. Amino Acid Squence: EVLQQDKSTSSNLHEAASD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| imp12, MTR10A, transportin 3, transportin-3 | |
| TNPO3 | |
| IgG | |
| Affinity Purified | |
| 96 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title