missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNPO3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76374-0.025mg
This item is not returnable.
View return policy
Description
TNPO3 Polyclonal specifically detects TNPO3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| TNPO3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| imp12, MTR10A, transportin 3, transportin-3 | |
| Rabbit | |
| 96 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| EAW83708 | |
| TNPO3 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human TNPO3. The immunogen is located within amino acids 220 - 270 of TNPO3. Amino Acid Squence: EVLQQDKSTSSNLHEAASD | |
| Affinity Purified | |
| RUO | |
| 23534 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction