missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
114.00 € - 375.00 €
Specifications
| Antigen | TIMP-3 |
|---|---|
| Dilution | Immunoprecipitation 1-5 uL/mg of lysate |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686744
|
Novus Biologicals
NBP3-21434-100ul |
100 μg |
375.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620684
|
Novus Biologicals
NBP3-21434-25ul |
25 μg |
114.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-3 Polyclonal antibody specifically detects TIMP-3 in Human samples. It is validated for ImmunoprecipitationSpecifications
| TIMP-3 | |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
| PBS, pH 7.0 | |
| 7078 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunoprecipitation 1-5 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Produced in rabbits immunized with E. coli-derived Human TIMP-3 fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title