missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21434-100ul
This item is not returnable.
View return policy
Description
TIMP-3 Polyclonal antibody specifically detects TIMP-3 in Human samples. It is validated for Immunoprecipitation
Specifications
| TIMP-3 | |
| Polyclonal | |
| Immunoprecipitation 1-5 uL/mg of lysate | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Produced in rabbits immunized with E. coli-derived Human TIMP-3 fragment | |
| 100 μg | |
| Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
| 7078 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| PBS, pH 7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction