Learn More
Tiabendazole, Mikromol™
Description
Tiabendazole
Specifications
Specifications
| Chemical Name or Material | Thiabendazole |
| CAS | 148-79-8 |
| Analyte or Component Names | Thiabendazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H7 N3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-(1,3-Thiazol-4-yl)-1H-benzimidazole, 2-(4-Thiazolyl)-1H-benzimidazole, 2-(4-Thiazolyl)-1H-benzoimidazole, 2-(4-Thiazolyl)benzimidazole, 2-(4-Thiazoyl)benzimidazole, 2-(4-Triazolyl)benzimidazole, 2-(4'-Thiazolyl)benzimidazole, 2-(4'-Thiazoyl)benzimidazole, 4-(1H-Benzimidazol-2-yl)thiazole, 5-(4-Thiazolyl)benzimidazole, Amolden HS, Bovizole, Chemviron TK 100, Cropasal, Drawipas, Eprofil, Equizole, G 491, Hokkustar HP, Hokustar HP, Irgaguard F 3000, Lombristop, MK 360, MSD 18, Marukacide M 101, Mertect, Mertect 160, Mertect 340F, Mertect LSP, Metasol TK 100, Metasol TK 25, Metasol TK 25AD, Metasol TK 50AD, Mintesol, Mintezol, Minzolum, NSC 525040, NSC 90507, Nemapan, Omnizole, Ormogal, Pitrizet, Polival, Sanaizol 100, Sanaizol TBZ-FL 25, Sistesan, Storite, Syntol M 100, TBZ, TBZ (fungicide), TBZ 6, TBZ 60W, TK 25AD, Tebuzate, Tecta, Tectab, Tecto, Tecto 10P, Tecto 40F, Tecto 60, Tecto B, Thiaben, Thiabendazol, Thiabendazole, Thiabendole, Thiabenzole, Thibendole, Thibenzol, Thibenzole, Thibenzole 200, Tiabenda, Tiabendazol, Tiabendazole, Tibimix 20, Top Form Wormer, Triasox |
| InChI Formula | InChI=1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13) |
| SMILES | c1ccc2[nH]c(nc2c1)c3cscn3 |
| IUPAC Name | 4-(1H-benzimidazol-2-yl)-1,3-thiazole |
| Show More |