Learn More
Tiabendazole, LoGiCal
Product Code. 30459710
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459710 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459710 Supplier LoGiCal Supplier No. LGCFOR0602.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thiabendazole
CAS 148-79-8
Analyte or Component Names Thiabendazole
Shipping Condition Room Temperature
Molecular Formula C10 H7 N3 S
Certifications/Compliance ISO 9001
Synonym 2-(1,3-Thiazol-4-yl)-1H-benzimidazole, 2-(4-Thiazolyl)-1H-benzimidazole, 2-(4-Thiazolyl)-1H-benzoimidazole, 2-(4-Thiazolyl)benzimidazole, 2-(4-Thiazoyl)benzimidazole, 2-(4-Triazolyl)benzimidazole, 2-(4'-Thiazolyl)benzimidazole, 2-(4'-Thiazoyl)benzimidazole, 4-(1H-Benzimidazol-2-yl)thiazole, 5-(4-Thiazolyl)benzimidazole, Amolden HS, Bovizole, Chemviron TK 100, Cropasal, Drawipas, Eprofil, Equizole, G 491, Hokkustar HP, Hokustar HP, Irgaguard F 3000, Lombristop, MK 360, MSD 18, Marukacide M 101, Mertect, Mertect 160, Mertect 340F, Mertect LSP, Metasol TK 100, Metasol TK 25, Metasol TK 25AD, Metasol TK 50AD, Mintesol, Mintezol, Minzolum, NSC 525040, NSC 90507, Nemapan, Omnizole, Ormogal, Pitrizet, Polival, Sanaizol 100, Sanaizol TBZ-FL 25, Sistesan, Storite, Syntol M 100, TBZ, TBZ (fungicide), TBZ 6, TBZ 60W, TK 25AD, Tebuzate, Tecta, Tectab, Tecto, Tecto 10P, Tecto 40F, Tecto 60, Tecto B, Thiaben, Thiabendazol, Thiabendazole, Thiabendole, Thiabenzole, Thibendole, Thibenzol, Thibenzole, Thibenzole 200, Tiabenda, Tiabendazol, Tiabendazole, Tibimix 20, Top Form Wormer, Triasox
InChI Formula InChI=1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)
Solution Type Neat
SMILES c1ccc2[nH]c(nc2c1)c3cscn3
IUPAC Name 4-(1H-benzimidazol-2-yl)-1,3-thiazole
Industry Type Clinical Testing
Molecular Weight (g/mol) 201.25
Quantity 10 mg
Formula Weight 201.0361 g/mol