Learn More
Thymol Blue solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12970554 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12970554 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12970554 Supplier Fisher Chemical Supplier No. RP 219 100
Please to purchase this item. Need a web account? Register with us today!
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Spezifikation
Packaging Bottle
Analyte or Component Names Thymol Blue ,
Purity Grade Notes EU Ph Ref: 1090601
Element Thymol Blue
Quantity 100 mL
Grade Eu. Ph.
compliance-icons
Produktidentifikator
  • Thymol blue solution. European Pharmacopoeia Ref: 1090601. (RP 219)
Signalwort
  • Warning
Gefahrenkategorie
  • Flammable liquid Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .