missing translation for 'onlineSavingsMsg'
Learn More

Thulium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

Catalog Number p-7083826
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7440-30-4
Tm
168.93
MFCD00011281
FRNOGLGSGLTDKL-UHFFFAOYSA-N
23961
CHEBI:33380
thulium
[Tm]
This item is not returnable. View return policy
7440-30-4
Tm
168.93
MFCD00011281
FRNOGLGSGLTDKL-UHFFFAOYSA-N
23961
CHEBI:33380
thulium
[Tm]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix HNO3
Chemical Name or Material Thulium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'