Learn More
Thieno[3,2-c]pyridine, Mikromol™
Product Code. 30052553
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052553 25 mg 25mg
30051011 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052553 Supplier Mikromol Supplier No. MM0150.120025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Thieno[3,2-c]pyridine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thieno[3,2-c]pyridine
CAS 272-14-0
Analyte or Component Names Thieno[3,2-c]pyridine
Shipping Condition Room Temperature
Molecular Formula C7 H5 N S
Certifications/Compliance ISO 9001
Synonym Ticlopidine Hydrochloride Imp. A (EP), Ticlopidine Imp. A (EP), Thieno[3,2-c]pyridine, Ticlopidine Hydrochloride Impurity A, Ticlopidine Impurity A
InChI Formula InChI=1S/C7H5NS/c1-3-8-5-6-2-4-9-7(1)6/h1-5H
SMILES c1cc2sccc2cn1
IUPAC Name thieno[3,2-c]pyridine
Molecular Weight (g/mol) 135.19
Quantity 25 mg
Formula Weight 135.0143 g/mol