Learn More
Thieno[3,2-c]pyridine, Mikromol™
Description
Thieno[3,2-c]pyridine
Specifications
Specifications
| Chemical Name or Material | Thieno[3,2-c]pyridine |
| CAS | 272-14-0 |
| Analyte or Component Names | Thieno[3,2-c]pyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H5 N S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ticlopidine Hydrochloride Imp. A (EP), Ticlopidine Imp. A (EP), Thieno[3,2-c]pyridine, Ticlopidine Hydrochloride Impurity A, Ticlopidine Impurity A |
| InChI Formula | InChI=1S/C7H5NS/c1-3-8-5-6-2-4-9-7(1)6/h1-5H |
| SMILES | c1cc2sccc2cn1 |
| IUPAC Name | thieno[3,2-c]pyridine |
| Show More |