Learn More
Thiamazole, LoGiCal
Product Code. 30458884
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458884 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458884 Supplier LoGiCal Supplier No. LGCFOR0889.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Methyl-2-(methylsulfanyl)-1H-imidazole
CAS 14486-52-3
Analyte or Component Names 1-Methyl-2-(methylsulfanyl)-1H-imidazole
Shipping Condition Room Temperature
Molecular Formula C5 H8 N2 S
Certifications/Compliance ISO 9001
Synonym 1-Methyl-2-(methylsulphanyl)-1H-imidazole, Thiamazole Imp. C (EP), 1H-Imidazole, 1-methyl-2-(methylthio)- (9CI), Imidazole, 1-methyl-2-(methylthio)- (8CI), 1-Methyl-2-(methylthio)imidazole, 2-Methylthio-3-methylimidazole, Methimazole USP Related Compound C, Methimazole USP RC C, Methimazole Related Compound C, Thiamazole Impurity C
InChI Formula InChI=1S/C5H8N2S/c1-7-4-3-6-5(7)8-2/h3-4H,1-2H3
Solution Type Neat
SMILES CSc1nccn1C
IUPAC Name 1-methyl-2-methylsulfanylimidazole
Industry Type Clinical Testing
Molecular Weight (g/mol) 128.2
Quantity 10 mg
Formula Weight 128.0408 g/mol