Learn More
Thiabendazole-5-hydroxy 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30321167
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321167 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30321167 Supplier Dr. Ehrenstorfer Supplier No. DREA17450500AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thiabendazole-5-hydroxy
CAS 948-71-0
Analyte or Component Names Thiabendazole-5-hydroxy
Shipping Condition +20°C
Molecular Formula C10 H7 N3 O S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1H-Benzimidazol-5-ol, 2-(4-thiazolyl)- (9CI), 5(or 6)-Benzimidazolol, 2-(4-thiazolyl)- (7CI), 5-Benzimidazolol, 2-(4-thiazolyl)- (8CI), 2-(4-Thiazolyl)-1H-benzimidazol-6-ol, 5-Hydroxythiabendazole
InChI Formula InChI=1S/C10H7N3OS/c14-6-1-2-7-8(3-6)13-10(12-7)9-4-15-5-11-9/h1-5,14H,(H,12,13)
SMILES Oc1ccc2[nH]c(nc2c1)c3cscn3
IUPAC Name 2-(1,3-thiazol-4-yl)-1H-benzimidazol-5-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 217.25
Quantity 1 mL
Formula Weight 217.031 g/mol