Learn More
Theophylline EP Impurity D (Diprophylline EP Impurity A), Mikromol™
Description
Theophylline EP Impurity D (Diprophylline EP Impurity A)
Specifications
Specifications
| Chemical Name or Material | N-Methyl-5-(methylamino)-1H-imidazole-4-carboxamide (Theophyllidine) |
| CAS | 6736-40-9 |
| Analyte or Component Names | N-Methyl-5-(methylamino)-1H-imidazole-4-carboxamide (Theophyllidine) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C6 H10 N4 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Diprophylline Imp. A (EP), Theophylline Imp. D (EP), Theophylline-Ethylenediamine Anhydrous Imp. D (EP), Theophylline USP Related Compound D, Theophylline USP RC D, N-Methyl-5-(methylamino)-1H-imidazole-4-carboxamide, Theophyllidine, Theophylline-Ethylenediamine Imp. D (EP), 1H-Imidazole-4-carboxamide, N-methyl-5-(methylamino)-, Imidazole-4(or 5)-carboxamide, N-methyl-5(or 4)-methylamino- (6CI), Imidazole-4-carboxamide, N-methyl-5-(methylamino)- (8CI), 4-(Methylamino)-5-(N-methylcarbamoyl)imidazole, Theophyllidine (imidazole derivative), Theophylline Related Compound D, Diprophylline Impurity A, Theophylline Impurity D, Theophylline-Ethylenediamine Impurity D |
| InChI Formula | InChI=1S/C6H10N4O/c1-7-5-4(6(11)8-2)9-3-10-5/h3,7H,1-2H3,(H,8,11)(H,9,10) |
| SMILES | CNC(=O)c1nc[nH]c1NC |
| IUPAC Name | N-methyl-5-(methylamino)-1H-imidazole-4-carboxamide |
| Show More |