Learn More
Thenalidine Tartrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Thenalidine Tartrate |
| CAS | 2784-55-6 |
| Analyte or Component Names | Thenalidine Tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H22 N2 S . C4 H6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Piperidinamine, 1-methyl-N-phenyl-N-(2-thienylmethyl)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), 2-Thenylamine, N-(1-methyl-4-piperidyl)-N-phenyl-, tartrate (5CI), 4-Piperidinamine, 1-methyl-N-phenyl-N-(2-thienylmethyl)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), Piperidine, 1-methyl-4-(N-2-thenylanilino)-, tartrate (1:1) (8CI), Piperidine, 1-methyl-4-(N-2-thenylanilino)-, tartrate (7CI), Tartaric acid, compd. with 1-methyl-4-N-2-thenylanilinopiperidine (6CI), 1-Methyl-4-(N-2-thenylanilino)piperidine tartrate, 1-Methyl-4-amino-N-phenyl-N-(2-thenyl)piperidine tartrate, AS 716, Sandostein, Sandosten, Thenalidine tartrate, Thenophenopiperidine tartrate |
| InChI Formula | InChI=1S/C17H22N2S.C4H6O6/c1-18-11-9-16(10-12-18)19(14-17-8-5-13-20-17)15-6-3-2-4-7-15;5-1(3(7)8)2(6)4(9)10/h2-8,13,16H,9-12,14H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1 |
| Solution Type | Neat |
| SMILES | CN1CCC(CC1)N(Cc2cccs2)c3ccccc3.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |