Learn More
Tetryzoline Hydrochloride, LoGiCal
Product Code. 30463624
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463624 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463624 Supplier LoGiCal Supplier No. LGCFOR0088.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetryzoline hydrochloride
CAS 522-48-5
Analyte or Component Names Tetryzoline hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H16 N2 . Cl H
Certifications/Compliance ISO 9001
Synonym Tetryzoline Hydrochloride, 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole hydrochloride (1:1), 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole monohydrochloride, 2-(1,2,3,4-Tetrahydro-1-naphthyl)-2-imidazoline monohydrochloride, 2-(1,2,3,4-Tetrahydro-1-naphthyl)-2-imidazoline hydrochloride, Murine Plus, Soothe, Tetrahydrozoline hydrochloride, Tetryzoline hydrochloride, Tinarhinin, Tizine, Tyzanol, Tyzanol hydrochloride, Tyzine, Tyzine hydrochloride, Visine, Visine hydrochloride, Yxin
InChI Formula InChI=1S/C13H16N2.ClH/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13;/h1-2,4,6,12H,3,5,7-9H2,(H,14,15);1H
Solution Type Neat
SMILES Cl.C1CC(C2=NCCN2)c3ccccc3C1
IUPAC Name 2-(1,2,3,4-tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazole;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 236.74
Quantity 10 mg
Formula Weight 236.108 g/mol