Learn More
Memmert™ Temperature Profile Write/Read Unit (Pc/Chip Card) Up To 40 Ramps, For Ctc256
Shop All Memmert Products
Click to view available options
Data Outputs:
Up to 40 Ramps
Unit Size:
Each
Description
- For programming via PC
- For writing to and reading from the chip card
- Up to 40 ramps
Specifications
Specifications
| Data Outputs | Up to 40 Ramps |
| Description | Up to 40 ramps, programming via PC |
| For Use With (Equipment) | Models 200, 400, 500, 108, 153, 246, 256 |
| Type | Temperature Profile Unit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto