missing translation for 'onlineSavingsMsg'
Learn More

TEA, 0.2M buffer soln., pH 8.5, Thermo Scientific™

Catalog Number 15464219
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
Triethanolamine buffer solution
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
This item is not returnable. View return policy
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
Triethanolamine buffer solution
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material TEA buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'