Learn More
Tamsulosin EP Impurity H (as Racemate Hydrochloride), Mikromol™
Description
Tamsulosin EP Impurity H (as Racemate Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride |
| CAS | 2724727-12-0 |
| Analyte or Component Names | (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H27 N O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydrochloride, Tamsulosin Hydrochloride Imp. H (EP), Tamsulosin Imp. H (EP), (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride, Tamsulosin Hydrochloride Imp. H (EP) as Racemate Hydrochloride, Tamsulosin Hydrochloride Impurity H as Racemate Hydrochloride, Tamsulosin Impurity H as Racemate Hydrochloride |
| InChI Formula | InChI=1S/C20H28N2.H3O4P/c1-16(2)22(17(3)4)15-13-19(18-10-6-5-7-11-18)20-12-8-9-14-21-20;1-5(2,3)4/h5-12,14,16-17,19H,13,15H2,1-4H3;(H3,1,2,3,4) |
| Solution Type | Neat |
| SMILES | Cl.CCOc1ccccc1OCCNC(C)Cc2ccc(OC)cc2 |
| Show More |