missing translation for 'onlineSavingsMsg'
Learn More

Tamsulosin EP Impurity H (as Racemate Hydrochloride), Mikromol™

Product Code. 17993806
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993806 100 mg 100mg
30049635 25 mg 25mg
30460222 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993806 Supplier Mikromol Supplier No. MM0938.01
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tamsulosin EP Impurity H (as Racemate Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride
CAS 2724727-12-0
Analyte or Component Names (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride
Shipping Condition Room Temperature
Molecular Formula C20 H27 N O3 . Cl H
Certifications/Compliance ISO 17025
Synonym (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydrochloride, Tamsulosin Hydrochloride Imp. H (EP), Tamsulosin Imp. H (EP), (2RS)-N-[2-(2-Ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine Hydro-chloride, Tamsulosin Hydrochloride Imp. H (EP) as Racemate Hydrochloride, Tamsulosin Hydrochloride Impurity H as Racemate Hydrochloride, Tamsulosin Impurity H as Racemate Hydrochloride
InChI Formula InChI=1S/C20H28N2.H3O4P/c1-16(2)22(17(3)4)15-13-19(18-10-6-5-7-11-18)20-12-8-9-14-21-20;1-5(2,3)4/h5-12,14,16-17,19H,13,15H2,1-4H3;(H3,1,2,3,4)
SMILES Cl.CCOc1ccccc1OCCNC(C)Cc2ccc(OC)cc2
IUPAC Name N-[2-(2-ethoxyphenoxy)ethyl]-1-(4-methoxyphenyl)propan-2-amine;hydrochloride
Molecular Weight (g/mol) 365.89
Quantity 100 mg
Formula Weight 365.1758 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.