Learn More
Tamsulosin EP Impurity A (as Hydrochloride), Mikromol™
Description
Tamsulosin EP Impurity A (as Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | 5-[(2R)-2-[Bis[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride |
| CAS | 2727060-83-3 |
| Analyte or Component Names | 5-[(2R)-2-[Bis[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H40 N2 O7 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-[(2R)-2-[Bis[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride, Tamsulosin Hydrochloride Imp. A (EP) as Hydrochloride, Tamsulosin Hydrochloride Impurity A as Hydrochloride, Tamsulosin Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C30H40N2O7S.ClH/c1-5-36-25-11-7-9-13-27(25)38-19-17-32(18-20-39-28-14-10-8-12-26(28)37-6-2)23(3)21-24-15-16-29(35-4)30(22-24)40(31,33)34;/h7-16,22-23H,5-6,17-21H2,1-4H3,(H2,31,33,34);1H/t23-;/m1./s1 |
| Solution Type | Neat |
| SMILES | Cl.CCOc1ccccc1OCCN(CCOc2ccccc2OCC)[C@H](C)Cc3ccc(OC)c(c3)S(=O)(=O)N |
| Show More |