missing translation for 'onlineSavingsMsg'
Learn More

Synthetic Fresh Water Standard 160-180 ppm, Reagecon

Product Code. 11583222 Shop All Reagecon Products
Click to view available options
Quantity:
5 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11583222

Brand: Reagecon HSH1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Can be used as a calibration, control, or method validation product, or as a known matrix in which to dissolve or dilute other products.

  • Water Reference Standard, Water Reference Material
  • Can be used as a calibration, control, or method validation product, or as a known matrix in which to dissolve or dilute other products
  • Specification presented is water hardness in ppm CaC03

Specifications

Packaging HDPE Container
Physical Form Liquid
Concentration 160-180 ppm
Quantity 5 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.