missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation SunFire C18 Column

Product Code. 13280340
Encompass
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Particle Size:
2.5 μm
3.5 μm
5 μm
10 μm
Unit Size:
Each
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
13280340 100 mm 2.1 mm 3.5 μm Each
13280360 150 mm 4.6 mm 3.5 μm Each
13270340 50 mm 2.1 mm 3.5 μm Each
13280350 150 mm 3 mm 3.5 μm Each
10667725 100 mm 4.6 mm 5 μm Each
13270350 100 mm 3 mm 3.5 μm Each
10236385 100 mm 3 mm 5 μm Each
13270360 100 mm 4.6 mm 3.5 μm Each
13210370 50 mm 4.6 mm 5 μm Each
13135079 30 mm 2.1 mm 2.5 μm Each
10563647 50 mm 2.1 mm 2.5 μm Each
13250360 50 mm 4.6 mm 3.5 μm Each
13165069 250 mm 4.6 mm 10 μm Each
13115089 30 mm 3 mm 2.5 μm Each
13135089 50 mm 3 mm 2.5 μm Each
12193142 50 mm 4.6 mm 2.5 μm Each
10696206 250 mm 4.6 mm 5 μm Each
13290340 150 mm 2.1 mm 3.5 μm Each
13230350 50 mm 2.1 mm 5 μm Each
13240350 100 mm 2.1 mm 5 μm Each
13250350 150 mm 2.1 mm 5 μm Each
13260350 50 mm 3 mm 3.5 μm Each
13290350 50 mm 3 mm 5 μm Each
13210360 150 mm 3 mm 5 μm Each
13220360 250 mm 3 mm 5 μm Each
13260360 75 mm 4.6 mm 3.5 μm Each
13200370 30 mm 4.6 mm 5 μm Each
13220370 150 mm 4.6 mm 5 μm Each
13155069 150 mm 4.6 mm 10 μm Each
17173222 75 mm 2.1 mm 2.5 μm Each
15838712 75 mm 3 mm 2.5 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
This item is not returnable. View return policy
Product Code. 13280340 Supplier Waters Corporation Supplier No. 186002534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SunFire C18 columns are used for general purpose method development at low pH.

  • The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 100 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.