missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, MedChemExpress

Product Code. 16448727
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16448727

Brand: Medchem Express HY14655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer.

Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer.

Specifications

Chemical Name or Material Sulfasalazine
CAS 599-79-1
Color Orange
Purity Grade Notes Research
Molecular Formula C18H14N4O5S
Quantity 5 g
Synonym NSC 667219
Solubility Information NH4OH : 150 mg/mL (376.52 mM; ultrasonic and adjust pH to 9 with NH4OH) ∣DMSO : 80 mg/mL (200.81 mM; Need ultrasonic and warming)
SMILES OC(C1=C(O)C=CC(/N=N/C2=CC=C(S(=O)(NC3=NC=CC=C3)=O)C=C2)=C1)=O
Molecular Weight (g/mol) 398.39
Formula Weight 398.39
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.