Learn More
Sulfapyridine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Sulfapyridine
Specifications
Specifications
| Chemical Name or Material | Sulfapyridine |
| CAS | 144-83-2 |
| Analyte or Component Names | Sulfapyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H11 N3 O2 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sulfasalazine Imp. J (EP), Benzenesulfonamide, 4-amino-N-2-pyridinyl-Sulfanilamide, N1-2(1H)-pyridylidene- (6CI,7CI), Sulfanilamide, N1-2-pyridyl- (8CI), Sulfanilamide, N4-2-pyridyl- (4CI), 2-Sulfanilamidopyridine, 2-Sulfapyridine, 4-Amino-N-2-pyridinylbenzenesulfonamide, 4-[(2-Pyridylamino)sulfonyl]aniline, A-499, Adiplon, Coccoclase, Dagenan, Eubasin, Eubasinum, Haptocil, M and B 693, N-(2-Pyridyl)-4-aminobenzenesulfonamide, N1-2-Pyridylsulfanilamide, NSC 41791, NSC 4753, Piridazol, Pirodazole, Plurazol, Pyriamid, Pyridazol, Relbapiridina, Ronin, Septipulmon, Streptosilpyridine, Sulfapyridine, Sulfidin, Sulfidine, Sulphapyridine, Thioseptal, Trianon, Sulfasalazine Impurity J |
| InChI Formula | InChI=1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14) |
| SMILES | Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2 |
| IUPAC Name | 4-amino-N-(2-pyridyl)benzenesulfonamide |
| Show More |