Learn More
Sulfapyridine 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326432
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30326432
Brand: Dr. Ehrenstorfer DREA17000100AL1000
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfapyridine
CAS 144-83-2
Analyte or Component Names Sulfapyridine
Shipping Condition +20°C
Molecular Formula C11 H11 N3 O2 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfasalazine Imp. J (EP), Benzenesulfonamide, 4-amino-N-2-pyridinyl-Sulfanilamide, N1-2(1H)-pyridylidene- (6CI,7CI), Sulfanilamide, N1-2-pyridyl- (8CI), Sulfanilamide, N4-2-pyridyl- (4CI), 2-Sulfanilamidopyridine, 2-Sulfapyridine, 4-Amino-N-2-pyridinylbenzenesulfonamide, 4-[(2-Pyridylamino)sulfonyl]aniline, A-499, Adiplon, Coccoclase, Dagenan, Eubasin, Eubasinum, Haptocil, M and B 693, N-(2-Pyridyl)-4-aminobenzenesulfonamide, N1-2-Pyridylsulfanilamide, NSC 41791, NSC 4753, Piridazol, Pirodazole, Plurazol, Pyriamid, Pyridazol, Relbapiridina, Ronin, Septipulmon, Streptosilpyridine, Sulfapyridine, Sulfidin, Sulfidine, Sulphapyridine, Thioseptal, Trianon, Sulfasalazine Impurity J
InChI Formula InChI=1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2
IUPAC Name 4-amino-N-(2-pyridyl)benzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 249.29
Quantity 1 mL
Formula Weight 249.288 g/mol