Learn More
Sulbactam EP Impurity F, Mikromol™
Product Code. 30462128
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462128 4 x 25 mg -
30051286 25 mg 25mg
30049435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30462128 Supplier Mikromol Supplier No. MM0308.0400254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sulbactam EP Impurity F, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CAS 24158-88-1
Analyte or Component Names (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Shipping Condition Room Temperature
Molecular Formula C8 H9 Br2 N O3 S
Certifications/Compliance ISO 9001
Synonym (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (6,6-Dibromopenicillanic Acid), Sulbactam Sodium Imp. F (EP), Sulbactam Imp. F (EP), Sulbactam USP Related Compound F, Sulbactam USP RC F, (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6,6-dibromo-3,3-dimethyl-7-oxo-, (2S-cis)- (8CI), 6,6-Dibromopenicillanic acid, 6,6-Dibromopenicillanic Acid, Sulbactam Related Compound F, Sulbactam Sodium Impurity F, Sulbactam Impurity F
InChI Formula InChI=1S/C8H9Br2NO3S/c1-7(2)3(4(12)13)11-5(14)8(9,10)6(11)15-7/h3,6H,1-2H3,(H,12,13)/t3-,6+/m0/s1
Solution Type Neat
SMILES CC1(C)S[C@H]2N([C@H]1C(=O)O)C(=O)C2(Br)Br
IUPAC Name (2S,5R)-6,6-dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 359.04
Quantity 4 x 25 mg
Formula Weight 356.867 g/mol