Learn More
Sulbactam EP Impurity F, Mikromol™
Description
Sulbactam EP Impurity F
Specifications
Specifications
| Chemical Name or Material | (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| CAS | 24158-88-1 |
| Analyte or Component Names | (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H9 Br2 N O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (6,6-Dibromopenicillanic Acid), Sulbactam Sodium Imp. F (EP), Sulbactam Imp. F (EP), Sulbactam USP Related Compound F, Sulbactam USP RC F, (2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6,6-dibromo-3,3-dimethyl-7-oxo-, (2S-cis)- (8CI), 6,6-Dibromopenicillanic acid, 6,6-Dibromopenicillanic Acid, Sulbactam Related Compound F, Sulbactam Sodium Impurity F, Sulbactam Impurity F |
| InChI Formula | InChI=1S/C8H9Br2NO3S/c1-7(2)3(4(12)13)11-5(14)8(9,10)6(11)15-7/h3,6H,1-2H3,(H,12,13)/t3-,6+/m0/s1 |
| SMILES | CC1(C)S[C@H]2N([C@H]1C(=O)O)C(=O)C2(Br)Br |
| IUPAC Name | (2S,5R)-6,6-dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Show More |