missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus Agalactiae Antibody (224/46), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62933-0.025MG
This item is not returnable.
View return policy
Description
Streptococcus Agalactiae Monoclonal specifically detects Streptococcus Agalactiae in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Streptococcus Agalactiae | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Streptococcus agalactiae (NCTC 11360) | |
| 0.025 mg | |
| Primary | |
| Bacteria | |
| IgG3 |
| ELISA | |
| 224/46 | |
| ELISA 1:100-1:2000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| This product is specific for Streptococcus agalactiae a gram-positive streptococcus characterized by the presence of the group B Lancefield antigen. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction