missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus Agalactiae Antibody (224/46), Novus Biologicals™
Mouse Monoclonal Antibody
212.00 € - 518.00 €
Specifications
| Antigen | Streptococcus Agalactiae |
|---|---|
| Clone | 224/46 |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084656
|
Novus Biologicals
NB100-62933-0.025MG |
0.025 mg |
212.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18019903
|
Novus Biologicals
NB100-62933 |
0.1 mg |
518.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Streptococcus Agalactiae Monoclonal specifically detects Streptococcus Agalactiae in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Streptococcus Agalactiae | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Bacteria | |
| IgG3 | |
| Affinity Purified | |
| This product is specific for Streptococcus agalactiae a gram-positive streptococcus characterized by the presence of the group B Lancefield antigen. |
| 224/46 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Streptococcus agalactiae (NCTC 11360) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title