missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Standard Format HE33 Mini Submarine Unit and Accessories

Product Code. 11390452 Shop All Cytiva Products
Click to view available options
:
HE 33 Mini Submarine Unit; Type: complete
This item is not returnable. View return policy

Product Code. 11390452

Brand: Cytiva 80605245

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Basic HE 33 Mini Submarine Unit:
Gel running tray, gel casting tray and bubble level

Complete HE 33 Mini Submarine Unit:
Basic unit, 1.5mm thick 8-well comb and comb back

Enhanced passive cooling system allows running gels at high voltages for speed and resolution

  • Coolant-insulated buffer chamber runs 7 x 10cm agarose gels in <20 minutes
  • Convenient, leak-proof casting trays provide fast gel casting without tape
  • UV-transparent gel tray for convenient gel handling and viewing
  • Combs allow up to 32 samples per run
  • Molded base design to be filled with common coolant such as 50% ethylene glycol

Order Info

For Basic unit, combs and comb back should be ordered separately

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Type Mini Submarine Unit (Complete)
Includes Basic unit, 1.5mm thick 8-well comb and comb back
Sample Size Up to 32
Temperature (English) Operating 122°F
Temperature (Metric) Operating 50°C
Volume (English) Buffer 8.4 oz.
Volume (Metric) Buffer 250 mL
Dimensions (L x W x H) 5.2 x 9.5 x 2.8 in. (13 x 24 x 7 cm)
Length (English) 5.2 in.
Length (English) Gel 3.9 in.
Length (Metric) 13 cm
Length (Metric) Gel 10 cm
Width (English) 9.5 in.
Width (English) Gel 2.7 in.
Width (Metric) 24 cm
Width (Metric) Gel 7 cm
Height (English) 2.8 in.
Height (Metric) 7 cm
Voltage 500 V
Wattage 15 w
Amperage 500 mA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.