Learn More
Cytiva Stainless Steel Staining Tray
Product Code. 10036134 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10036134 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10036134 Supplier Cytiva Supplier No. 80634524
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
For Use With (Application) For gel staining
Type Staining Tray
Includes Gasket, Splash Guard, Integral Magnet Plate, Tubing
Material Stainless Steel
Quantity 1 Each