Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for IN110, 15 mm
Product Code. 17507365 Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17507365
Brand: Memmert™ IN110B02073
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) IN110 Incubator
Material Stainless Steel
Type Drip Tray
Height (Metric) Rim: 15 mm