missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPON2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | SPON2 |
|---|---|
| Concentration | 0.41 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870055
|
Proteintech
20513-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860055
|
Proteintech
20513-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).Specifications
| SPON2 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BUD6 | |
| 10417 | |
| SPON2 Fusion Protein Ag14337 | |
| Primary | |
| -20°C | |
| SPON2 |
| 0.41 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DIL 1, DIL1, M spondin, Mindin, SPON2, Spondin 2 | |
| SPON2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title