Learn More
SPON2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20513-1-AP-150UL
 View more versions of this product
Product Code. 16860055
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).
Specifications
Specifications
SPON2
Polyclonal
Unconjugated
SPON2
DIL 1, DIL1, M spondin, Mindin, SPON2, Spondin 2
Rabbit
Antigen Affinity Chromatography
RUO
10417
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BUD6
SPON2
SPON2 Fusion Protein Ag14337
150 μL