missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
205.00 € - 490.00 €
Specifications
| Antigen | SOX2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry Assay dependent, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence Assay dependent, Immunohistochemistry-Paraffin |
| Applications | Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251172
|
Novus Biologicals
NB110-79875SS |
0.025 mg |
205.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757263
|
Novus Biologicals
NB110-79875 |
490.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Rat, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SOX2 | |
| Western Blot 1:100-1:2000, Flow Cytometry Assay dependent, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence Assay dependent, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| PBS with 0.02% Sodium Azide | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| SOX2 | |
| IgG | |
| Affinity Purified | |
| SOX-2 |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Rat, Human, Mouse, Fish | |
| P48431 | |
| 6657 | |
| Reacts with residues 113-127 of the 37 kDa (predicted molecular weight is 34 kDa) human, mouse and rat SOX-2 protein. Sequence is 100% conserved in most species. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 34.3 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title