missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB110-79875SS
This item is not returnable.
View return policy
Description
SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Rat, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SOX2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Rabbit | |
| 34.3 kDa | |
| 0.025 mg | |
| Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 6657 | |
| Rat, Human, Mouse, Fish | |
| IgG |
| Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry Assay dependent, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence Assay dependent, Immunohistochemistry-Paraffin | |
| P48431 | |
| SOX2 | |
| Reacts with residues 113-127 of the 37 kDa (predicted molecular weight is 34 kDa) human, mouse and rat SOX-2 protein. Sequence is 100% conserved in most species. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| SOX-2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction