Learn More
Sorafenib, Mikromol™
Description
Sorafenib
Specifications
Specifications
| Chemical Name or Material | Sorafenib |
| CAS | 284461-73-0 |
| Analyte or Component Names | Sorafenib |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H16 Cl F3 N4 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, BAY 43-9006, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N'-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, Nevaxar, Sorafenib |
| InChI Formula | InChI=1S/C21H16ClF3N4O3/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25/h2-11H,1H3,(H,26,30)(H2,28,29,31) |
| SMILES | CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)ccn1 |
| IUPAC Name | 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-methylpyridine-2-carboxamide |
| Show More |