Learn More
Sodium phosphate, 0.2M buffer soln., pH 9.5, Thermo Scientific™
Product Code. 15455419
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Product Code. 15455419
Brand: Thermo Scientific Alfa Aesar J63159.AP
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.2 M
pH 9.5
Physical Form Liquid
Quantity 500 mL
MDL Number MFCD00146206