missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 1M buffer soln., pH 8.5

Product Code. 15493179
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 15493179

Brand: Thermo Scientific Alfa Aesar J60408.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 1 L
Chemical Name or Material Sodium bicarbonate buffer soln.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.