Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 100.3 cSt (40°C), Paragon Scientific™
Product Code. 16070170
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
Product Code. 16070170
Brand: Paragon Scientific HVS06
Please to purchase this item. Need a web account? Register with us today!
Certified in strict accordance with ASTM D2162 at 40 °C and 100 °C under our ISO 17025 and ISO 17034 accreditation and manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids.
  • Fully traceable to international standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Certified in strict accordance with ASTM D2162
  • 60mL lower volume cost effective solution
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications
Quantity 60 mL
Nominal values quoted. Certified Values will vary from batch to batch.