Learn More
Sitafloxacin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Sitafloxacin |
| CAS | 127254-12-0 |
| Analyte or Component Names | Sitafloxacin |
| Shipping Condition | +20°C |
| Molecular Formula | C19 H18 Cl F2 N3 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-Quinolinecarboxylic acid, 7-[(7S)-7-amino-5-azaspiro[2.4]hept-5-yl]-8-chloro-6-fluoro-1-[(1R,2S)-2-fluorocyclopropyl]-1,4-dihydro-4-oxo-, 3-Quinolinecarboxylic acid, 7-(7-amino-5-azaspiro[2.4]hept-5-yl)-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-1,4-dihydro-4-oxo-, [1R-[1α(S*),2α]]-, 5-Azaspiro[2.4]heptane, 3-quinolinecarboxylic acid deriv., 7-[(7S)-7-Amino-5-azaspiro[2.4]hept-5-yl]-8-chloro-6-fluoro-1-[(1R,2S)-2-fluorocyclopropyl]-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, DU 6859, DU 6859a, Gracevit; |
| InChI Formula | InChI=1S/C19H18ClF2N3O3/c20-14-15-8(17(26)9(18(27)28)5-25(15)12-4-10(12)21)3-11(22)16(14)24-6-13(23)19(7-24)1-2-19/h3,5,10,12-13H,1-2,4,6-7,23H2,(H,27,28)/t10-,12+,13+/m0/s1 |
| SMILES | N[C@@H]1CN(CC12CC2)c3c(F)cc4C(=O)C(=CN([C@@H]5C[C@@H]5F)c4c3Cl)C(=O)O |
| Show More |