Learn More
Silver Nitrate 0.05 M (0.05 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA
Product Code. 11552942 Shop All Reagecon Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
Product Code. 11552942
Brand: Reagecon N20050W
Please to purchase this item. Need a web account? Register with us today!
Fully factorized, high purity, certified, stable product, developed and tested for precipitation (argentometric) titrations.
  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified by an ISO17025 Accredited Test Method (INAB Ref. 264T)
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Oxidation/reduction titrations depend on changes in the electropotential of the oxidation reduction system and always involves a reaction between an oxidizing and reducing pair.
Specifications
Description Accuracy ± 0.1%
Packaging Amber Glass Winchester Bottle GL45
Physical Form Liquid
Concentration 0.05 M
Technique Titration
Quantity 2.5 L
compliance-icons
Product Identifier
  • silver nitrate 0.05M
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : silver nitrate