Learn More
Sibutramine EP Impurity B, Mikromol™
Description
Sibutramine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | (1RS)-1-(1-(3-Chlorophenyl)cyclobutyl)-N,N,3-trimethylbutan-1-amine Hydrochloride |
| CAS | 2727060-96-8 |
| Analyte or Component Names | (1RS)-1-(1-(3-Chlorophenyl)cyclobutyl)-N,N,3-trimethylbutan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 Cl N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS)-1-[1-(3-Chlorophenyl)cyclobutyl]-N,N,3-trimethylbutan-1-amine Hydrochloride, Sibutramine Hydrochloride Monohydrate Imp. B (Pharmeuropa) as Hydrochloride, Sibutramine Related Compound B as Hydrochloride, Sibutramine Hydrochloride Monohydrate Impurity B, Sibutramine Impurity B |
| InChI Formula | InChI=1S/C17H26ClN.ClH/c1-13(2)11-16(19(3)4)17(9-6-10-17)14-7-5-8-15(18)12-14;/h5,7-8,12-13,16H,6,9-11H2,1-4H3;1H |
| SMILES | Cl.CC(C)CC(N(C)C)C1(CCC1)c2cccc(Cl)c2 |
| IUPAC Name | 1-[1-(3-chlorophenyl)cyclobutyl]-N,N,3-trimethylbutan-1-amine;hydrochloride |
| Show More |