missing translation for 'onlineSavingsMsg'
Learn More

SERCA2 ATPase Antibody (IID8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications

218.00 €

Specifications

Antigen SERCA2 ATPase
Clone IID8
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:5000, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:500, Block/Neutralize
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18548890
View Documents
Novus Biologicals
NB300-529-0.025ml
0.025 mL
218.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications

Specifications

SERCA2 ATPase
1 mg/mL
Western Blot
Unconjugated
RUO
O46674
488
Purified canine cardiac sarcoplasmic reticulum.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
IID8
Western Blot 1:500 - 1:5000, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:500, Block/Neutralize
Monoclonal
Mouse
Human
ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2
ATP2A2
IgG1
Affinity Purified
Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.