Learn More
SERCA2 ATPase Antibody (IID8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-529-0.025ml
This item is not returnable.
View return policy
Description
SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
| SERCA2 ATPase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:5000, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:500, Block/Neutralize | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 488 | |
| Human | |
| IgG1 |
| Western Blot | |
| IID8 | |
| Unconjugated | |
| O46674 | |
| ATP2A2 | |
| Purified canine cardiac sarcoplasmic reticulum. | |
| 0.025 mL | |
| Primary | |
| Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms. | |
| Store at -20°C. Avoid freeze/thaw cycles. |