Learn More
Se4+ solution for ICP 100mg/L, Fisher Chemical™
BIOPHARMA
Product Code. 12980774 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12980774 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12980774 Supplier Fisher Chemical Supplier No. SP 004 100
Please to purchase this item. Need a web account? Register with us today!
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix 2% HNO3
Quantity 100 mL
Chemical Name or Material Se4+ solution