missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 258719, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
SB 258719 is a selective 5-HT7 receptor antagonist with high affinity (pKi=7.5) for the receptor. SB 258719 can be used for the research of cancer and neurological disease.
Specifications
Specifications
| Chemical Name or Material | SB 258719 |
| CAS | 195199-95-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H30N2O2S |
| Quantity | 25 mg |
| Solubility Information | DMSO : 250 mg/mL (738.53 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC=CC(C)=C1)(N(C)[C@H](C)CCN2CCC(C)CC2)=O |
| Molecular Weight (g/mol) | 338.51 |
| Formula Weight | 338.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction