Learn More
Sacubitril Sodium, Mikromol
Description
Sacubitril Sodium, Mikromol
Specifications
Specifications
| Chemical Name or Material | Sacubitril Sodium |
| CAS | 149690-05-1 |
| Analyte or Component Names | Sacubitril Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H28 N O5 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, sodium salt (1:1), (αR,γS)-, [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, monosodium salt, [S-(R*,S*)]- (9CI), Sac |
| InChI Formula | InChI=1S/C24H29NO5.Na/c1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19;/h4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28);/q;+1/p-1/t17-,21+;/m1./s1 |
| Solution Type | Neat |
| SMILES | [Na+].CCOC(=O)[C@H](C)C[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)CCC(=O)[O-] |
| Show More |