missing translation for 'onlineSavingsMsg'
Learn More

S6K2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen S6K2
Concentration 0.19 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16874364
View Documents
Proteintech
15268-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16864364
View Documents
Proteintech
15268-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains a kinase catalytic domain and phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications

Specifications

S6K2
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Mouse, Rat, Human
Q9UBS0, Q9Z1M4
361696, 58988, 6199
S6K2 Fusion Protein Ag7371
Primary
-20°C
RPS6KB2
0.19 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KLS, P70 beta, P70 beta 1, P70 beta 2, p70 ribosomal S6 kinase beta, P70 S6 kinase beta, p70 S6K 2, p70 S6K beta, p70 S6KB, p70(S6K) beta, P70S6K2, p70S6Kb, RPS6KB2, S6 kinase related kinase, S6K beta, S6K beta 2, S6K beta2, S6K2, SRK, STK14B
Rps6kb2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.